Learn More
Chem-Lab 100ML Colour stock standard sol. 500 A.P.H.A.units (1.246 g K2PtCl6 + 1 g CoCl2.6H2O + 10
Brand: Chem-Lab CL01.0860.0100
100ML Colour stock standard sol. 500 A.P.H.A.units (1.246 g K2PtCl6 + 1 g CoCl2.6H2O + 10
This item is not returnable.
View return policy
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid, Cobalt(II) chloride,Potassium hexachloroplatinate(IV)