missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lubricating Oil Standard 1: Ca @ 0.000; P @ 0.000; S @ 0.000; Zn @ 0.000 wt% in Lubricating Oil, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Lubricating Oil Standard 1: Ca @ 0.000; P @ 0.000; S @ 0.000; Zn @ 0.000 wt% in Lubricating Oil
Specifications
Specifications
| Chemical Name or Material | Cadmium/Phosphorus/Sulfur/Zinc |
| CAS | 7440-43-9, 7723-14-0, 7704-34-9, 7440-66-6 |
| Methods | XRF |
| Analyte or Component Names | Cadmium/Phosphorus/Sulfur/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 20 cSt Mineral Oil |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Cadmium 0%/Phosphorus 0%/Sulfur 0%/Zinc 0% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction