missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-5/Siglec-14 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Description
Siglec-5/Siglec-14 Monoclonal specifically detects Siglec-5/Siglec-14 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | Siglec-5/Siglec-14 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 194128 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | O15389 |
| Gene Symbols | SIGLEC5 |
| Show More |
For Research Use Only (RUO)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?