missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgG1, kappa Recombinant Isotype Control (5Vk2/37.7H)
Human Recombinant Monoclonal Isotype Control
Brand: Invitrogen™ MA555090
This item is not returnable.
View return policy
Description
Human IgG1, kappa Isotype Control for Flow
Human IgG1, also known as human immunoglobulin G1, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the most abundant subclass of IgG in the blood and plays a key role in defense against pathogens. IgG1 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG1 is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG1 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Specifications
| Human IgG1, kappa | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| 100 μg | |
| Not Applicable | |
| Isotype Control | |
| IgG1 κ |
| Flow Cytometry | |
| 5Vk2/37.7H | |
| Unconjugated | |
| IGHG1; immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Protein A/G | |
| RUO | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction