missing translation for 'onlineSavingsMsg'
Learn More

PAMCI Antibody (OTI5E5), Novus Biologicals Biologicals™

Product Code. 30306128 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30306128 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30306128 Supplier Novus Biologicals Supplier No. NBP344747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PAMCI Monoclonal antibody specifically detects PAMCI in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAMCI
Applications Western Blot
Classification Monoclonal
Clone OTI5E5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation Lyophilized powder ( 1X PBS, pH 7.3,)
Gene Alias PAM COOH-terminal interactor protein 1, PAMCI, PCIP1, P-CIP1peptidylglycine alpha-amidating monooxygenase COOH-terminal interactorprotein-1, Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor, Ras association (RalGDS/AF-6) domain family (N-terminal) member 9, ras association domain-containing protein 9
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-232 of human RASSF9 (NP_005438) produced in E.coli.
Purification Method >95%
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9182
Reconstitution For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.