missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L1 Antibody (5E2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42929
This item is not returnable.
View return policy
Description
Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Apolipoprotein L1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 500 | |
| Western Blot 1:10000 - 1:50000, ELISA, Immunohistochemistry 1:50 - 1:500 | |
| ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4 | |
| apolipoprotein L, 1 - (Uniprot# O14791) | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| 8542 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction