missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytoglobin Antibody (7F6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42955
This item is not returnable.
View return policy
Description
Cytoglobin Monoclonal antibody specifically detects Cytoglobin in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Cytoglobin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 114757 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 7F6 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| cytoglobin, HGBHistoglobin, histoglobin, STAPHGb, Stellate cell activation-associated protein | |
| cytoglobin - (Uniprot# Q8WWM9) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction