missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGBP1 Antibody (6E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42985
This item is not returnable.
View return policy
Description
IGBP1 Monoclonal antibody specifically detects IGBP1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| IGBP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 6000 | |
| Western Blot 1:200 - 1:1000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16 | |
| immunoglobulin(CD79A) binding protein 1 - (Uniprot# P78318) | |
| 100 μg | |
| Protein Phosphatase, Signal Transduction | |
| 3476 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction