missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTF2 Antibody (5C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43008
This item is not returnable.
View return policy
Description
NTF2 Monoclonal antibody specifically detects NTF2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| NTF2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 5C7 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:250 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| NTF-2, NTF2PP15Placental protein 15, nuclear transport factor 2 | |
| nuclear transport factor 2 - (Uniprot# P61970) | |
| 100 μg | |
| Signal Transduction | |
| 10204 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction