Learn More
Invitrogen™ CDK2 Recombinant Rabbit Monoclonal Antibody (24GB4315)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557367
 View more versions of this product
Product Code. 30276293
  • € 564.00 / 100µL
Description
CDK2 Recombinant Monoclonal Antibody for Western Blot, Flow
CDK2/cyclinO is a member of the cyclin dependent protein kinase family complexed to CyclinO. CyclinO was used as a specific protein isoform name for the UNG gene which is localized exclusively in the nucleus. In addition, CyclinO was reported to shows significant uracil-DNA glycosylase activity. CDK2/cyclinO protein levels increase during G1 phase and the protein is turned over during the course of cell cycle. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDK2
Recombinant Monoclonal
Unconjugated
CDK2
A630093N05Rik; CDC28; CDC2A; cdc2-related protein kinase; CDK1; CDK2; CDKN2; Cell division protein kinase 2; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 2; EC 2.7.11.22; EC 2.7.11.23; kinase Cdc2; MPF; p33 protein kinase; p33(CDK2); p33CDK2; p34 protein kinase
Rabbit
Affinity chromatography
RUO
1017, 12566, 362817
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4315
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P24941, P97377, Q63699
CDK2
A synthesized peptide derived from human Cdk2 (aa 281-298).
100 μL
Primary
Human, Mouse, Rat