missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chikungunya Virus NSP4 (S27-African prototype) Recombinant Rabbit Monoclonal Antibody (HL2580)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556647
This item is not returnable.
View return policy
Description
Chikungunya Virus NSP4 (S27-African prototype) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Chikungunya fever is a pandemic disease caused by the mosquito-borne Chikungunya virus (CHIKV). The viral enzyme non-structural protein 4 (nsP4), as RNA-dependent RNA polymerase (RdRP), catalyzes the formation of negative-sense, genomic and subgenomic viral RNAs.
Specifications
| Chikungunya Virus NSP4 (S27-African prototype) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2580 | |
| Unconjugated | |
| Chikungunya Virus | |
| Recombinant fragment of Chikungunya virus nsP4 (strain S27-African prototype). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction