Learn More
Invitrogen™ Ferritin Heavy Chain Recombinant Rabbit Monoclonal Antibody (HL1905)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556706
This item is not returnable.
View return policy
Description
Ferritin Heavy Chain Recombinant Monoclonal Antibody for Western Blot
FTH1 is the heavy subunit of ferritin, the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in ferritin proteins are associated with several neurodegenerative diseases. This gene has multiple pseudogenes. Several alternatively spliced transcript variants have been observed, but their biological validity has not been determined.
Specifications
| Ferritin Heavy Chain | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P02794, Q2MHN2, Q95MP7 | |
| FTH1 | |
| Recombinant fragment of human Ferritin Heavy Chain. | |
| 100 μL | |
| Primary | |
| Human, Canine, Feline | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1905 | |
| Unconjugated | |
| FTH1 | |
| apoferritin; cell proliferation-inducing gene 15 protein; Ferritin H subunit; ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain, N-terminally processed; Ferritin subunit H; ferritin, heavy polypeptide 1; FHC; frih; FTH; Fth1; FTHL6; HFE5; H-ferritin; HFt; MFH; MGC104426; OK/SW-cl.84; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100499480, 2495, 403631, 654516 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |