missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXA1 Recombinant Rabbit Monoclonal Antibody (HL1975)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556713
This item is not returnable.
View return policy
Description
FOXA1 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
Specifications
| FOXA1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P55317 | |
| FOXA1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FOXA1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1975 | |
| Unconjugated | |
| FOXA1 | |
| Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; hepatocyte nuclear factor 3-alpha; HNF3A; HNF-3A; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3169 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction