missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSIG1 Recombinant Rabbit Monoclonal Antibody (0Q2B5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555473
This item is not returnable.
View return policy
Description
Immunogen Sequence: ASAKLDFANN VQLSLTLAAL SLGLWWTFDR SRSGLGLGIT IAFLATLITQ FLVYNGVYQY TSPDFLYIRS WLPCIFFSGG VTVGNIGRQL AMGVPEKPHS D.
Oxysterols regulate cholesterol homeostasis through liver X receptor (LXR) and sterol regulatory element-binding protein (SREBP) mediated signaling pathway. This gene is an insulin-induced gene. It encodes an endoplasmic reticulum (ER) membrane protein that plays a critical role in regulating cholesterol concentrations in cells. This protein binds to the sterol-sensing domains of SREBP cleavage-activating protein (SCAP) and HMG CoA reductase, and is essential for the sterol-mediated trafficking of the two proteins. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| INSIG1 | |
| Recombinant Monoclonal | |
| 1.19 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q08755, Q8BGI3 | |
| INSIG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 177-277 of human INSIG1. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 0Q2B5 | |
| Unconjugated | |
| INSIG1 | |
| 1810013C12Rik; CL6; CL-6; fb55a03; fi35b10; growth response protein (CL-6); immediate-early protein CL-6; INSIG1; INSIG-1; INSIG-1 membrane protein; insulin induced gene 1; insulin induced protein 1; insulin-induced gene 1 protein; Insulin-induced growth response protein CL-6; MGC1405; wu:fb55a03; wu:fi35b10; zgc:55439 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 231070, 64194 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction