Learn More
Invitrogen™ MBD2 Recombinant Rabbit Monoclonal Antibody (24GB4570)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557474
 View more versions of this product
Product Code. 30275766
  • € 564.00 / 100µL
Description
MBD2 Recombinant Monoclonal Antibody for Western Blot, Flow
Heterochromatin protein-1 (HP1) is a methyl-lysine binding protein localized at heterochromatin sites, where it mediates gene silencing. It has been shown that mammalian methyltransferases that selectively methylate histone H3 on lysine-9 generate a binding site for HP1 proteins, a family of heterochromatic adaptor molecules implicated in both gene silencing and supranucleosomal chromatin structure. High-affinity in vitro recognition of a methylated histone H3 peptide by HP1 requires a functional chromodomain. Thus, the HP1 chromodomain is a specific interaction motif for the methyl epitope on lysine-9 of histone H3. In vivo, heterochromatin association of HP1 proteins is lost in Suv39h double-null primary mouse fibroblasts but is restored after reintroduction of a catalytically active SUV39H1 HMTase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MBD2
Recombinant Monoclonal
Unconjugated
Mbd2
demethylase; DKFZp586O0821; DMTase; MBD2; MBD2a; methyl-CpG binding domain protein 2; methyl-CpG-binding domain protein 2; Methyl-CpG-binding protein MBD2; NY-CO-41
Rabbit
Affinity chromatography
RUO
17191, 680172, 8932
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4570
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UBB5, Q9Z2E1
Mbd2
A synthesized peptide derived from human MBD2 (aa 216-234).
100 μL
Primary
Human, Mouse, Rat