missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma pneumoniae P1 (strain ATCC 29342/M129/Subtype 1) Recombinant Rabbit Monoclonal Antibody (HL3204)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556805
This item is not returnable.
View return policy
Description
Mycoplasma pneumoniae P1 (strain ATCC 29342/M129/Subtype 1) Recombinant Monoclonal Antibody for ELISA, LF
Although they are the simplest and smallest self replicating bacteria, Mycoplasma infections tend to be persistent, difficult to detect, diagnose, and cure. Mycoplasmas are surface parasites of human respiratory and urogenital tracts. Mycoplasma pneumoniae is often the causative agent of community-acquired pneumonia. The affinity of the organism for respiratory epithelial cells and its ability to secrete hydrogen peroxide are a potent combination, leading to oxidative tissue damage.
Specifications
| Mycoplasma pneumoniae P1 (strain ATCC 29342/M129/Subtype 1) | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA, Lateral Flow | |
| HL3204 | |
| Unconjugated | |
| M. pneumoniae | |
| Recombinant fragment of Mycoplasma pneumoniae P1 protein.(strain# ATCC 29342/M129/Subtype 1). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction