missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43369
This item is not returnable.
View return policy
Description
Alkaline Phosphatase Polyclonal antibody specifically detects Alkaline Phosphatase in Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Alkaline Phosphatase | |
| Polyclonal | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Immunized with recombinant Mouse Alkaline Phosphatase (Uniprot#: P24823 & P09242; Phe18∽Arg335) | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 0.5-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction