missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDA R, NR1 Subunit Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43401
This item is not returnable.
View return policy
Description
NMDA R, NR1 Subunit Polyclonal antibody specifically detects NMDA R, NR1 Subunit in Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Serum,Immunocytochemistry/ Immunofluorescence
Specifications
| NMDA R, NR1 Subunit | |
| Polyclonal | |
| Unconjugated | |
| PBS, pH 7.4, 50% glycerol | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 0.2-2 μg/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation, Serum | |
| GluN1, MRD8, NDHMSD, NDHMSR;NMD-R1, NMDA1, NMDAR1, NR1 | |
| Immunized with recombinant Rat NMDA R, NR1 Subunit (Uniprot#: P35439; Arg19∽Gln559) | |
| 100 μL | |
| Cellular Signaling, Neuroscience | |
| 2902 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction