Learn More
Invitrogen™ RbAp48 Recombinant Rabbit Monoclonal Antibody (24GB1240)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557680
 View more versions of this product
Product Code. 30276495
  • € 564.00 / 100µL
Description
RbAp48 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RbAp48-11G10 recognizes full length RbAp48, a 48 kDa nuclear protein first isolated by its ability to interact with the carboxyl-terminus of the retinoblastoma protein (Rb). It is a member of the WD (Trp-Asp) repeat family of proteins. RbAp48 is one of the three subunits of CAF-1, can bind to histone H4 and is a subunit of human histone deacetylase HD1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RbAp48
Recombinant Monoclonal
Unconjugated
RBBP4
CAF-1 p48 subunit; CAF-1 subunit C; CAF-I 48 kDa subunit; CAF-I p48; chromatin assembly factor 1 subunit C; Chromatin assembly factor I p48 subunit; chromatin assembly factor/CAF-1 p48 subunit; histone-binding protein RBBP4; lin-53; mRbAp48; MSI1 protein homolog; nucleosome-remodeling factor subunit RBAP48; NURF55; RB binding protein 4, chromatin remodeling factor; RBAP48; RBBP4; RBBP-4; retinoblastoma binding protein 4; retinoblastoma-binding protein 4; Retinoblastoma-binding protein p48
Rabbit
Affinity chromatography
RUO
19646, 313048, 5928
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1240
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q09028, Q60972
RBBP4
A synthesized peptide derived from human RbAp48 (aa 2-51).
100 μL
Primary
Human, Mouse, Rat