missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETD1A Recombinant Rabbit Monoclonal Antibody (HL2705)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556871
This item is not returnable.
View return policy
Description
SETD1A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IP
SET1A is a component of a histone methyltransferase (HMT) complex that produces mono-, di-, and trimethylated histone H3 at Lys4. The complex is the analog of the S. cerevisiae Set1/COMPASS complex.
Specifications
| SETD1A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O15047 | |
| SETD1A | |
| Recombinant fragment of human SETD1A. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| HL2705 | |
| Unconjugated | |
| SETD1A | |
| BC010250; Histone-lysine N-methyltransferase SETD1A; hSET1A; KIAA0339; KMT2F; Lysine N-methyltransferase 2F; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain containing 1A; SET domain-containing protein 1A; SET1; Set1/Ash2 histone methyltransferase complex subunit SET1; SET1A; SETD1A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 233904, 9739 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction