Learn More
Keystone Cleanroom Products 1073B Tyvek™ Autoclaveable Wrapper Large
BIOPHARMAGREENER_CHOICE
Product Code. 17798457 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
30 x 24 in.
30 x 30 in.
36 x 36 in.
40 x 40 in.
54 x 28 in.
54 x 54 in.
60 x 60 in.
80 x 40 in.
140 x 60 in.
Unit Size:
Pack of 100
Pack of 250
Product Code. 17798457
Brand: Keystone Cleanroom Products MGTVKSHEET2854
Only null left
Add to basket
Add to basket
Provides an excellent bacterial barrier for superior performance post steam sterilization. Cut using an automated CAD system in ISO 8 and made from DuPont 1073B Medical Grade Tyvek.
  • Designed for Steam Sterilization
  • Recommended for Use in Conjunction with a Secondary Tyvek™ Sterilization Pouch
  • Hydrophobic
  • Excellent Microbial Barrier
  • Resists Tearing and Punctures
  • Lint-Free - Category (Helmke Drum)
  • Manufactured and Packaged in a ISO 6 Cleanroom
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type None
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Sterilization Method Steam
Temperature Range (English) <250°F
Temperature Range (Metric) <125°C
Dimensions (L x W) 54 x 28 in.
Length (English) 54 in.
Length (Metric) 137.2 cm
Width (English) 28 in.
Width (Metric) 71.2 cm
Product Type Sterilization Wrap