missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenol, TRC

Product Code. 18083220
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083220 10 g 10g
30137353 100 g 100g
30136887 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083220 Supplier Toronto Research Chemicals Supplier No. TRCD46585510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-26-1
Molecular Formula C8 H10 O
Molecular Weight (g/mol) 122.16
Synonym 2,6-Xylenol (8CI), 2,6-Dimethylphenol, 1-Hydroxy-2,6-dimethylbenzene, D 104339, NSC 2123
IUPAC Name 2,6-dimethylphenol
SMILES Cc1cccc(C)c1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H10O/c1-6-4-3-5-7(2)8(6)9/h3-5,9H,1-2H3
Formula Weight 122.0732
Chemical Name or Material 2,6-Dimethylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.