missing translation for 'onlineSavingsMsg'
Learn More

10H-Pyrido[3,2-b][1,4]benzothiazine, TRC

Product Code. 30423726
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423726 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423726 Supplier Toronto Research Chemicals Supplier No. TRCP991655250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Azaphenothiazine
CAS 261-96-1
Molecular Formula C11 H8 N2 S
Quantity 250 mg
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine, 1H-Pyrido[3,2-b][1,4]benzothiazine (9CI), 1-Azaphenothiazine, NSC 277720
InChI Formula InChI=1S/C11H8N2S/c1-2-5-9-8(4-1)13-11-10(14-9)6-3-7-12-11/h1-7H,(H,12,13)
SMILES N1c2ccccc2Sc3cccnc13
IUPAC Name 10H-pyrido[3,2-b][1,4]benzothiazine
Molecular Weight (g/mol) 200.26
Formula Weight 200.0408
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.