missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Deoxycortisol, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
11-Deoxycortisol
Specifications
Specifications
| Chemical Name or Material | 11-Deoxycortisol |
| CAS | 152-58-9 |
| Analyte or Component Names | 11-Deoxycortisol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H30 O4 |
| Synonym | Hydrocortisone Imp. F (EP), 17,21-Dihydroxypregn-4-ene-3,20-dione, Reichstein's Substance S, 11-Deoxycortisol, Cortexolone, Hydrocortisone Impurity F |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | C[C@]12 CCC(=O)C=C1CC[C@@H]3 [C@@H]2 CC[C@@]4(C)[C@H]3 CC[C@]4(O)C(=O)CO |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction