missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-epi-Testosterone, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Epitestosterone |
| CAS | 481-30-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H28 O2 |
| Quantity | 10 mg |
| Synonym | 17alpha-Hydroxyandrost-4-en-3-one, Testosterone Imp. C (EP), Androst-4-en-3-one','17-hydroxy-','(17α)-, Androst-4-en-3-one','17α-hydroxy- (8CI), (17α)-17-Hydroxyandrost-4-en-3-one, 17-Epitestosterone, 17α-Hydroxyandrost-4-en-3-one, 17α-Hydroxyandrost-4-ene-3-one, 17α-Testosterone, 17α-cis-Testosterone, 4-Androstene-17α-ol-3-one, Androst-4-en-17α-ol-3-one, Epitestosterone, Isotestosterone, NSC 26499, Testosterone','cis-, cis-Testosterone, epi-Testosterone |
| InChI Formula | InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17+,18-,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@H]2O |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 288.42 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction