Learn More
3,5,3',5'-Tetraiodo Thyromandelic Acid (>85%), TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 3,5,3',5'-Tetraiodo Thyromandelic acid |
| CAS | 93647-48-4 |
| Molecular Formula | C14 H8 I4 O5 |
| Quantity | 10 mg |
| Synonym | Benzeneacetic acid','α-hydroxy-4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, acid','4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, 2-Hydroxy-2-[4-(4-hydroxy-3,5-diiodo-phenoxy)-3,5-diiodo-phenyl]acetic Acid, 3,5,3',5'-Tetraiodo Thyromandelic Acid, Mandelic acid','4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo- (7CI), α-Hydroxy-4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzeneacetic acid |
| InChI Formula | InChI=1S/C14H8I4O5/c15-7-3-6(4-8(16)12(7)20)23-13-9(17)1-5(2-10(13)18)11(19)14(21)22/h1-4,11,19-20H,(H,21,22) |
| SMILES | OC(C(=O)O)c1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1 |
| IUPAC Name | 2-hydroxy-2-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetic acid |
| Molecular Weight (g/mol) | 763.83 |
| Formula Weight | 763.655 |