Learn More
Amisulpride-d5 N-Oxide, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Amisulpride-d5 N-Oxide |
| CAS | 1794756-15-2 |
| Molecular Formula | C17H22D5N3O5S |
| Quantity | 10 mg |
| Synonym | 4-Amino-N-[(1-(ethyl-d5)-1-oxido-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxybenzamide, 4-Amino-N-[(1-(ethyl-d5)-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2- methoxybenzamide N-Oxide |
| InChI Formula | InChI=1S/C17H27N3O5S/c1-4-20(22)8-6-7-12(20)11-19-17(21)13-9-16(26(23,24)5-2)14(18)10-15(13)25-3/h9-10,12H,4-8,11,18H2,1-3H3,(H,19,21)/i1D3,4D2 |
| SMILES | O=C(C1=C(OC)C=C(N)C(S(=O)(CC)=O)=C1)NCC2CCCN2(C([2H])([2H])C([2H])([2H])[2H])=O |
| IUPAC Name | 4-amino-5-ethylsulfonyl-2-methoxy-N-[[1-oxido-1-(1,1,2,2,2-pentadeuterioethyl)pyrrolidin-1-ium-2-yl]methyl]benzamide |
| Molecular Weight (g/mol) | 390.51 |