missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azithromycin-d3, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Azithromycin-d3 |
| CAS | 163921-65-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C38 2H3 H69 N2 O12 |
| Quantity | 10 mg |
| Synonym | Azithromycin D3 |
| InChI Formula | InChI=1S/C38H72N2O12/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28/h20-33,35,41-43,45-46H,15-19H2,1-14H3/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1/i13D3 |
| SMILES | [2H]C([2H])([2H])N1C[C@H](C)C[C@@](C)(O)[C@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@@H](C)[C@H](O[C@H]3C[C@@](C)(OC)[C@@H](O)[C@H](C)O3)[C@@H](C)C(=O)O[C@H](CC)[C@@](C)(O)[C@H](O)[C@H]1C |
| IUPAC Name | (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,8,10,12,14-hexamethyl-6-(trideuteriomethyl)-1-oxa-6-azacyclopentadecan-15-one |
| Molecular Weight (g/mol) | 752 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction