missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) Phthalate-d4, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-2-ethylhexyl ester D4 |
| CAS | 93951-87-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 2H4 H34 O4 |
| Quantity | 10 mg |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, bis(2-ethylhexyl) ester (9CI), 1,2-Bis(2-ethylhexyl) 1,2-benzene-3,4,5,6-d4-dicarboxylate, Phthalic Acid, Bis(2-ethylhexyl) Ester D4 (phenyl D4) |
| InChI Formula | InChI=1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3/i11D,12D,15D,16D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OCC(CC)CCCC)c(C(=O)OCC(CC)CCCC)c1[2H] |
| IUPAC Name | bis(2-ethylhexyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 394.58 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction