Learn More
Enzo Life Sciences 10MG Dimethyl-bisphenol A
Brand:  Enzo Life Sciences BML-GR339-0010
Encompass_Preferred
10MG Dimethyl-bisphenol A
Product Code. 15145058
  • € 128.26 / 10mg
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Dimethyl-bisphenol A
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.