Learn More
Enzo Life Sciences 10mg DL-ALPHA-Difluoromethylornithine .hydrochloride . monohydrate
Brand:  Enzo Life Sciences ALX-270-283-M010
Encompass_Preferred
10mg DL-ALPHA-Difluoromethylornithine .hydrochloride . monohydrate
Product Code. 15128337
  • € 60.50 / 10mg
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • DL-alpha-Difluoromethylornithine . hydrochloride . monohydrate
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.