Learn More
Ethyl 3-oxo-2-Phenylpropanoate (>90%), TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-oxo-2-Phenylpropanoate |
| CAS | 17838-69-6 |
| Molecular Formula | C11H12O3 |
| Quantity | 10 mg |
| Synonym | Ipratropium Bromide Impurity 2 |
| InChI Formula | InChI=1S/C11H12O3/c1-2-14-11(13)10(8-12)9-6-4-3-5-7-9/h3-8,10H,2H2,1H3 |
| SMILES | CCOC(=O)C(C=O)c1ccccc1 |
| IUPAC Name | ethyl 3-oxo-2-phenylpropanoate |
| Molecular Weight (g/mol) | 192.21 |
| Formula Weight | 192.0786 |