missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ T2AA
GREENER_CHOICE

Product Code. 18786648
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18786648

Brand: Tocris Bioscience™ 4723/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Proliferating cell nuclear antigen (PCNA) inhibitor

T2AA is a proliferating cell nuclear antigen (PCNA) inhibitor; inhibits PCNA/PIP-box peptide interaction (IC50 ∽1 μM). Causes DNA replication stress by stalling DNA replication forks. Inhibits PCNA interaction with DNA polymerase δ and arrests cell growth in S-phase. Suppresses cancer cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (βS)-β-Amino-4-(4-hydroxyphenoxy)-3,5-diiodobenzenpropanol
CAS 1380782-27-3
Quantity 10 mg
Molecular Formula C15H15I2NO3
Formula Weight 511.09
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.