missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine-d12, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Triethanolamine-d12 |
| CAS | 1469758-99-3 |
| Molecular Formula | C6H3D12NO3 |
| Quantity | 10 mg |
| Synonym | 2,2',2''-Nitrilotris-ethan-1,1,2,2-d4-ol |
| InChI Formula | InChI=1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2 |
| SMILES | OC([2H])([2H])C([2H])([2H])N(C([2H])([2H])C([2H])([2H])O)C([2H])([2H])C([2H])([2H])O |
| Molecular Weight (g/mol) | 161.26 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction