missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valproic Acid-d6 Beta-D-Glucuronide, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Valproic Acid-d6 beta-D-Glucuronide |
| Molecular Formula | C14H18D6O8 |
| Quantity | 10 mg |
| Synonym | 1-(2-Propylpentanoate-d6) β-D-Glucopyranuronic Acid, 1-O-(Valproyl-d6)-β-D-glucopyranuronic Acid, Dipropylacetate-d6 Glucuronide, VPA-G-d6, Valproate-d6 Glucuronide, Valproic Acid-d6 Glucuronide |
| InChI Formula | InChI=1S/C14H24O8/c1-3-5-7(6-4-2)13(20)22-14-10(17)8(15)9(16)11(21-14)12(18)19/h7-11,14-17H,3-6H2,1-2H3,(H,18,19)/t8-,9-,10+,11-,14-/m0/s1/i1D3,2D3 |
| SMILES | O[C@H]([C@@H](C(O)=O)O[C@@H](OC(C(CCC([2H])([2H])[2H])CCC([2H])([2H])[2H])=O)[C@@H]1O)[C@@H]1O |
| Molecular Weight (g/mol) | 326.37 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction