missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM D5453 Sulfur and Nitrogen Kit: S @ 10 mg/kg in each level and N from Pyridine @ 0, 10, 100, 200, 300 mg/kg in Isooctane, VHG™
Description
ASTM D5453 Sulfur and Nitrogen Kit: S @ 10 mg/kg in each level and N from Pyridine @ 0, 10, 100, 200, 300 mg/kg in Isooctane
Specifications
Specifications
| Chemical Name or Material | Sulfur/Nitrogen |
| CAS | 7704-34-9, 7727-37-9 |
| Analyte or Component Names | Sulfur/Nitrogen |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Isooctane |
| Quantity | 5 x 2 mL |
| Concentration or Composition (by Analyte or Components) | Sulfur 10 mg/kg/Nitrogen 0 - 10 - 100 - 200 - 300 mg/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?