Learn More
11-Deoxy Fusidic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 11-Deoxyfusidic Acid |
| CAS | 1013937-16-0 |
| Molecular Formula | C31 H48 O5 |
| Quantity | 10 mg |
| Synonym | 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3-hydroxy-, (3α,4α,8α,9β,13α,14β,16β,17Z)-, (3α,4α,8α,9β,13α,14β,16β,17Z)-16-(Acetyloxy)-3-hydroxy-29-nordammara-17(20),24-dien-21-oic acid, ent-(17Z)-16α-(Acetyloxy)-3β-hydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-17(20),24-dien-21-oic acid, 11-Deoxyfusidic acid |
| InChI Formula | InChI=1S/C31H48O5/c1-18(2)9-8-10-21(28(34)35)27-23-11-12-26-29(5)15-14-24(33)19(3)22(29)13-16-30(26,6)31(23,7)17-25(27)36-20(4)32/h9,19,22-26,33H,8,10-17H2,1-7H3,(H,34,35)/b27-21-/t19-,22-,23-,24+,25-,26-,29-,30-,31-/m0/s1 |
| SMILES | C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2CC[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C |
| IUPAC Name | (2Z)-2-[(3R,4S,5S,8S,9S,10S,13R,14S,16S)-16-acetyloxy-3-hydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid |
| Molecular Weight (g/mol) | 500.7098 |
| Formula Weight | 500.3502 |