Learn More
11-Ethoxycarbonyldodecanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 11-Ethoxycarbonyldodecanoic Acid |
| CAS | 66003-63-2 |
| Molecular Formula | C14 H26 O4 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C14H26O4/c1-2-18-14(17)12-10-8-6-4-3-5-7-9-11-13(15)16/h2-12H2,1H3,(H,15,16) |
| SMILES | CCOC(=O)CCCCCCCCCCC(=O)O |
| IUPAC Name | 12-ethoxy-12-oxododecanoic acid |
| Molecular Weight (g/mol) | 258.35 |
| Formula Weight | 258.1831 |