Learn More
11-Methyloleoside, TRC
Product Code. 30482750
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482750 100 mg 100mg
30483161 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30482750 Supplier Toronto Research Chemicals Supplier No. TRCM324705100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60539-23-3
Molecular Formula C17 H24 O11
Molecular Weight (g/mol) 404.37
Synonym 2H-Pyran-4-acetic acid, 3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-, (2S,3E,4S)-, 2H-Pyran-4-acetic acid, 3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-, [2S-(2α,3E,4β)]-, 11-Methyloleoside, Methyloleoside, Oleoside 11-methyl ester
IUPAC Name 2-[(2S,3E,4S)-3-ethylidene-5-methoxycarbonyl-2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4H-pyran-4-yl]acetic acid
SMILES COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)\[C@@H]1CC(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C17H24O11/c1-3-7-8(4-11(19)20)9(15(24)25-2)6-26-16(7)28-17-14(23)13(22)12(21)10(5-18)27-17/h3,6,8,10,12-14,16-18,21-23H,4-5H2,1-2H3,(H,19,20)/b7-3+/t8-,10+,12+,13-,14+,16-,17-/m0/s1
Formula Weight 404.1319
Chemical Name or Material Elenolic acid glucoside