missing translation for 'onlineSavingsMsg'
Learn More

(11Beta,16Alpha,17Alpha)-9-Chloro-17-(2,2-dioxido-5H-1,2-oxathiol-4-yl)-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-androsta-1,4-dien-3-one Mometasone Furoate Impurity B, TRC

Product Code. 30358258
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358258 1 mg 1mg
30351531 10 mg 10mg
30350268 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358258 Supplier Toronto Research Chemicals Supplier No. TRCC3665151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223776-49-6
Molecular Formula C28 H31 Cl O8 S
Molecular Weight (g/mol) 563.06
Synonym Androsta-1,4-dien-3-one, 9-chloro-17-(2,2-dioxido-5H-1,2-oxathiol-4-yl)-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-, (11β,16α,17α)-, (11β,16α,17α)-9-Chloro-17-(2,2-dioxido-5H-1,2-oxathiol-4-yl)-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylandrosta-1,4-dien-3-one, Mometasone Furoate Imp. B (EP)
IUPAC Name [(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-17-(2,2-dioxo-5H-oxathiol-4-yl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C6=CS(=O)(=O)OC6

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C28H31ClO8S/c1-16-11-21-20-7-6-17-12-19(30)8-9-25(17,2)27(20,29)23(31)13-26(21,3)28(16,18-14-36-38(33,34)15-18)37-24(32)22-5-4-10-35-22/h4-5,8-10,12,15-16,20-21,23,31H,6-7,11,13-14H2,1-3H3/t16-,20+,21+,23+,25+,26+,27+,28+/m1/s1
Formula Weight 562.1428
Chemical Name or Material 4-[9-Chloro-17-[(furan-2-ylcarbonyl)oxy]-11beta-hydroxy-16alpha-methyl-3-oxoandrosta-1,4-dien-17beta-yl]-5H-1,2-oxathiole 2,2-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.