missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione 21-Acetate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione 21-Acetate
Specifications
Specifications
| Chemical Name or Material | 11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione 21-Acetate |
| CAS | 21940-45-4 |
| Analyte or Component Names | 11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione 21-Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H30 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pregna-4,6-diene-3,20-dione, 21-(acetyloxy)-11,17-dihydroxy-, (11β)- (9CI), Pregna-4,6-diene-3,20-dione, 11β,17,21-trihydroxy-, 21-acetate (6CI,7CI,8CI), Δ6-Cortisol 21-acetate, 11β,17,21-Trihydroxypregna-4,6-diene-3,20-dione 21-Acetate |
| InChI Formula | InChI=1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h4-5,10,16-18,20,26,28H,6-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1 |
| SMILES | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3C=CC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C |
| IUPAC Name | [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction