Learn More
11MM ELECTRONIC DECAPPER
Brand:  Fisherbrand
1154.25 EUR valid until 2024-12-31
Use promo code "23262" to get your promotional price.
11MM ELECTRONIC DECAPPER
Additional Details : Weight : 1.45500kg
Product Code. 16436180
  • € 1215.00 / Each
Estimated Shipment: 23-05-2024