missing translation for 'onlineSavingsMsg'
Learn More
Learn More
12-Lipoxygenase Antibody (OTI1C3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70165AF350
This item is not returnable.
View return policy
Description
12-Lipoxygenase Monoclonal specifically detects 12-Lipoxygenase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| 12-Lipoxygenase | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ALOX12 | |
| Full length recombinant protein of human 12-Lipoxygenase (NP_000688) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1C3 | |
| Western Blot, Immunohistochemistry | |
| 12(S)-lipoxygenase, 12-LOX, 12S-lipoxygenase, 12S-LOXarachidonate 12-lipoxygenase, 12S-type, arachidonate 12-lipoxygenase, EC 1.13.11, EC 1.13.11.31, LOG12, platelet 12-LOX, platelet-type 12-lipoxygenase, Platelet-type lipoxygenase 12 | |
| Mouse | |
| 0.1 mL | |
| 239 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction