missing translation for 'onlineSavingsMsg'
Learn More
Learn More
12-Lipoxygenase Antibody (OTI1C3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70165APC
This item is not returnable.
View return policy
Description
12-Lipoxygenase Monoclonal antibody specifically detects 12-Lipoxygenase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| 12-Lipoxygenase | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1C3 | |
| Western Blot, Immunohistochemistry | |
| 12(S)-lipoxygenase, 12-LOX, 12S-lipoxygenase, 12S-LOXarachidonate 12-lipoxygenase, 12S-type, arachidonate 12-lipoxygenase, EC 1.13.11, EC 1.13.11.31, LOG12, platelet 12-LOX, platelet-type 12-lipoxygenase, Platelet-type lipoxygenase 12 | |
| Full length human recombinant protein of human ALOX12 (NP_000688) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 239 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction