Learn More
Helios Italquartz 1234 DIN Standard Covers with Ring Handle, Transparent
Click to view available options
For Use With (Equipment):
CR 41-61 Crucible
CR 42-51-62 Crucible
CR 43-52-63 Crucible
CR 44-53-64 Crucible
CR 45-54-65 Crucible
CR 46-55-66 Crucible
CR 47-56 Crucible
Unit Size:
Each
Description
- Transparent
- Include ring handle
- Conform to 1234 DIN standard
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Specifications
| Color | Transparent |
| Material | Quartz |
| Type | 1234 DIN Standard Covers with Ring Handle |
| For Use With (Equipment) | CR 45-54-65 Crucible |