Learn More
Sartorius™ 13106 Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters
Brand: Sartorius™ 13106--47----HEN
Additional Details : Weight : 0.10000kg
Specifications
White | |
Cellulose Nitrate | |
0.45 μm | |
Used for colony counting, particle testing and microscopy | |
100 Pack | |
Membrane Filter | |
47 mm |
Membrane Filter | |
Cellulose Nitrate | |
Sterile | |
Black | |
115 to 145 μm | |
47 mm |
Safety and Handling
- X100 MBN NC EDGE W/B 0,45µ 47MM ST
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose