missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ 13806 Series Non-sterile Cellulose Acetate (CA) Membrane Filters

Brand:  Sartorius™ 13806--47------N

Additional Details : Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II

Product Code. 10169230

  • € 128.00 / Pack of 100
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Green
Cellulose Nitrate
0.45 μm
Used for colony counting, particle testing and microscopy
100 Pack
47 mm
Membrane Filter
Cellulose Nitrate
Non-sterile
Dark Green
Membrane Filter
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers