Learn More
14-3-3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97724MFV500
Product Code. 18596481
  • € 428.00 / 0.10mL
Description
14-3-3 Polyclonal antibody specifically detects 14-3-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mFluor Violet 500 SE
Produced in rabbits immunized with E. coli-derived Human 14-3-3 fragment.
Primary
Store at 4C in the dark.