Learn More
14-3-3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97724MFV500
This item is not returnable.
View return policy
Description
14-3-3 Polyclonal antibody specifically detects 14-3-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| mFluor Violet 500 SE | |
| Produced in rabbits immunized with E. coli-derived Human 14-3-3 fragment. | |
| Primary | |
| Store at 4C in the dark. |