missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-27202

 View more versions of this product

Product Code. 18719733

  • € 498.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

14-3-3 Polyclonal specifically detects 14-3-3 in Human, Mouse, Rat, Porcine, Canine, Chicken, Chinese Hamster, Feline, Opossum, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

14-3-3
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Rabbit
Protein A purified
RUO
Primary
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, panda, dog, Chinese hamster, chicken, opossum, platypus, pig, frog, cat, orangutan, crab-eating macaque, and rhesus monkey, 94% in Carolina anole, cow, zebrafish, and 89% homologous in frog, and rat.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
0.5 mg/mL
Western Blot 2-4ug/ml, Simple Western 1:50
P27348
YWHAQ
A portion of amino acids 125-175 from human 14-3-3 was used as the immunogen for this antibody.
0.1 mg
Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction
10971
Human, Mouse, Rat, Pig, Canine, Chicken, Chinese Hamster, Feline, Opossum, Other, Primate
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only