missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 beta Antibody (05), Biotin , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06157B
This item is not returnable.
View return policy
Description
14-3-3 beta Monoclonal antibody specifically detects 14-3-3 beta in Cynomolgus Monkey samples. It is validated for Western Blot, ELISA
Specifications
| 14-3-3 beta | |
| Monoclonal | |
| Biotin | |
| 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus 14-3-3 beta (Q4R572-2; Met 2-Asn 244). | |
| 0.1 mL | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
| Western Blot, ELISA | |
| 05 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 7529 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction